Learn More
Corning™ 1536-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 10082343 Shop All Corning Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
Product Code. Color unitSize
10082343 White Pack of 50
1 options
Product Code. 10082343 Supplier Corning™ Supplier No. 3727BC
Color:White; Sterility:Sterile; Lid:With Lid
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
  • Recommended working volume up to 8 μL
  • Round well bottom for reduced air entrapment and improved CV values and Z factor
  • Raised well bottom for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Format Standard
Lid With Lid
Culture Area 0.03 cm2
Array 32 x 48
Color White
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 12.5 μL
Well Shape Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color White