missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corn Oil, TRC
Specifications
Specifications
| Chemical Name or Material | Corn Oil |
| CAS | 8001-30-7 |
| Quantity | 500 mg |
| Synonym | Bunge, Corn Germ Oil, Germ Oil of Semen Maydis, Gold-N-Sweet Pan Spray, Lipomul, Maydol, Maise Oil, Mazola Oil, Semen Maydis Oil, Semen Maydis Salad Oil, Uni Ace CS |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?