missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CORIN Polyclonal Antibody

CORIN Polyclonal Antibody
Brand: Invitrogen PA542916
This item is not returnable.
View return policy
Description
CORIN Polyclonal Antibody for Western Blot
Specifications
| CORIN | |
| Polyclonal | |
| Unconjugated | |
| CORIN | |
| ATC2; Atrial natriuretic peptide-converting enzyme; Atrial natriuretic peptide-converting enzyme, 100 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 160 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 180 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, activated protease fragment; Atrial natriuretic peptide-converting enzyme, N-terminal propeptide; AV273130; Corin; corin, serine peptidase; CRN; EC 3.4.21; Heart-specific serine proteinase ATC2; low density lipoprotein receptor-related protein 4; low density lipoprotein-related protein 4; Lrp4; MGC119742; PEE5; pro-ANP-convertase; pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; TMPRSS10; Transmembrane protease serine 10 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10699 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y5Q5 | |
| CORIN | |
| Synthetic peptide directed towards the C-terminal of human CORIN (aa 881-930). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction