missing translation for 'onlineSavingsMsg'
Learn More

Corin, MedChemExpress

Product Code. 16467117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467117 5 mg 5mg
16444398 10 mg 10mg
16440717 50 mg 50mg
16405296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467117 Supplier Medchem Express Supplier No. HY111048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1.

Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1.

Specifications

Chemical Name or Material Corin
Physical Form Solid
CAS 1808113-09-8
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C26H28N4O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (233.36 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C([C@H]2[C@H](N)C2)C=C1)CCCC3=CC=C(C(NC4=CC=CC=C4N)=O)C=C3
Molecular Weight (g/mol) 428.53
Formula Weight 428.53
Percent Purity 95.34%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.