missing translation for 'onlineSavingsMsg'
Learn More

Corin, MedChemExpress

Product Code. 16444398
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444398

Brand: Medchem Express HY111048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1.

Corin is a dual inhibitor of histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1.

Specifications

Chemical Name or Material Corin
Physical Form Solid
CAS 1808113-09-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H28N4O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (233.36 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C([C@H]2[C@H](N)C2)C=C1)CCCC3=CC=C(C(NC4=CC=CC=C4N)=O)C=C3
Molecular Weight (g/mol) 428.53
Formula Weight 428.53
Percent Purity 95.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.