missing translation for 'onlineSavingsMsg'
Learn More

Corin Antibody (MM0199-9H21), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18402956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402956 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402956 Supplier Novus Biologicals Supplier No. NBP212205JF646

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

Corin Monoclonal antibody specifically detects Corin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Corin
Applications Western Blot
Classification Monoclonal
Clone MM0199-9H21
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Host Species Rat
Immunogen Human recombinant Corin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10699
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.