missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corin Antibody (MM0199-9H21), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12205AF750
This item is not returnable.
View return policy
Description
Corin Monoclonal specifically detects Corin in Human samples. It is validated for Western Blot.
Specifications
| Corin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CORIN | |
| Human recombinant Corin | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot, Immunocytochemistry | |
| MM0199-9H21 | |
| Western Blot, Immunocytochemistry | |
| atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10 | |
| Rat | |
| 0.1 mL | |
| 10699 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction