missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Corey Lactone, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Corey Lactone 5-(4-Phenylbenzoate) |
| CAS | 31752-99-5 |
| Molecular Formula | C21 H20 O5 |
| Quantity | 1 g |
| Synonym | [1,1'-Biphenyl]-4-carboxylic acid, (3aR,4S,5R,6aS)-hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl ester, 4-Biphenylcarboxylic acid, 5-ester with 3,3aα,4,5,6,6aα-hexahydro-5β-hydroxy-4α-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)- (8CI), [1,1'-Biphenyl]-4-carboxylic acid, hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl ester, [3aR-(3aα,4α,5β,6aα)]-, 2H-Cyclopenta[b]furan, [1,1'-biphenyl]-4-carboxylic acid deriv., (-)-Corey lactone, Corey lactone |
| InChI Formula | InChI=1S/C21H20O5/c22-12-17-16-10-20(23)25-18(16)11-19(17)26-21(24)15-8-6-14(7-9-15)13-4-2-1-3-5-13/h1-9,16-19,22H,10-12H2/t16-,17-,18+,19-/m1/s1 |
| SMILES | OC[C@H]1[C@@H](C[C@@H]2OC(=O)C[C@H]12)OC(=O)c3ccc(cc3)c4ccccc4 |
| IUPAC Name | [(3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-5-yl] 4-phenylbenzoate |
| Molecular Weight (g/mol) | 352.38 |
| Formula Weight | 352.1311 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?