missing translation for 'onlineSavingsMsg'
Learn More

(+)-Corey Lactone, TRC

Product Code. 30393080
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393080 10 g
30400773 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393080 Supplier Toronto Research Chemicals Supplier No. TRCH29401010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39265-57-1
Molecular Formula C21 H20 O5
Molecular Weight (g/mol) 352.38
Synonym [3aS-(3aα,4α,5β,6aα)]-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl Ester [1,1'-Biphenyl]-4-carboxylic Acid, 2H-Cyclopenta[b]furan, [1,1'-Biphenyl]-4-carboxylic Acid Deriv., (3aS,4R,5S,6aR)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl Ester [1,1'-Biphenyl]-4-carboxylic Acid, 4-Phenylbenzoate Alcohol
IUPAC Name [(3aS,4R,5S,6aR)-4-(hydroxymethyl)-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-5-yl] 4-phenylbenzoate
SMILES OC[C@@H]1[C@H](C[C@H]2OC(=O)C[C@@H]12)OC(=O)c3ccc(cc3)c4ccccc4

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H20O5/c22-12-17-16-10-20(23)25-18(16)11-19(17)26-21(24)15-8-6-14(7-9-15)13-4-2-1-3-5-13/h1-9,16-19,22H,10-12H2/t16-,17-,18+,19-/m0/s1
Formula Weight 352.1311
Chemical Name or Material (+)-Corey Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.