missing translation for 'onlineSavingsMsg'
Learn More

(-)-Corey Lactone, TRC

Product Code. 30356918
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356918 Supplier Toronto Research Chemicals Supplier No. TRCC6950005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Corey Lactone 5-(4-Phenylbenzoate)
CAS 31752-99-5
Molecular Formula C21 H20 O5
Quantity 5 g
Synonym [1,1'-Biphenyl]-4-carboxylic acid, (3aR,4S,5R,6aS)-hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl ester, 4-Biphenylcarboxylic acid, 5-ester with 3,3aα,4,5,6,6aα-hexahydro-5β-hydroxy-4α-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)- (8CI), [1,1'-Biphenyl]-4-carboxylic acid, hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl ester, [3aR-(3aα,4α,5β,6aα)]-, 2H-Cyclopenta[b]furan, [1,1'-biphenyl]-4-carboxylic acid deriv., (-)-Corey lactone, Corey lactone
InChI Formula InChI=1S/C21H20O5/c22-12-17-16-10-20(23)25-18(16)11-19(17)26-21(24)15-8-6-14(7-9-15)13-4-2-1-3-5-13/h1-9,16-19,22H,10-12H2/t16-,17-,18+,19-/m1/s1
SMILES OC[C@H]1[C@@H](C[C@@H]2OC(=O)C[C@H]12)OC(=O)c3ccc(cc3)c4ccccc4
IUPAC Name [(3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-5-yl] 4-phenylbenzoate
Molecular Weight (g/mol) 352.38
Formula Weight 352.1311
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.