missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histone Macro-H2A.1 Monoclonal Antibody (CL5245)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531412
This item is not returnable.
View return policy
Description
Immunogen sequence: GEVSKAASAD STTEGTPA Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 94%.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| Histone Macro-H2A.1 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75367, Q02874, Q9QZQ8 | |
| MACROH2A1 | |
| Synthetic peptide corresponding to Human H2AFY. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL5245 | |
| Unconjugated | |
| MACROH2A1 | |
| Core histone macro-H2A.1; H2A histone family member Y; H2A histone family, member Y; H2A.y; H2A/y; H2AF12M; H2AFY; Histone H2A.y; histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; macroH2A1; MACROH2A1.1; macroH2A1.2; medulloblastoma antigen MU-MB-50.205; mH2A1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26914, 29384, 9555 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction