missing translation for 'onlineSavingsMsg'
Learn More

Cordycepin, MedChemExpress

Product Code. 16484797
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Product Code. Quantity unitSize
16484797 50 mg 50mg
16473846 25 mg 25mg
16418928 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16484797

Brand: Medchem Express HYN0262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cordycepin (3'-Deoxyadenosine) is a nucleoside derivative and inhibits IL-1β-induced MMP-1 and MMP-3 expression in rheumatoid arthritis synovial fibroblasts (RASFs) in a dose-dependent manner. Cordycepin kills Mycobacterium tuberculosis through hijacking the bacterial adenosine kinase.

Cordycepin (3'-Deoxyadenosine) is a nucleoside derivative and inhibits IL-1β-induced MMP-1 and MMP-3 expression in rheumatoid arthritis synovial fibroblasts (RASFs) in a dose-dependent manner. Cordycepin kills Mycobacterium tuberculosis through hijacking the bacterial adenosine kinase.

Specifications

Chemical Name or Material Cordycepin
CAS 73-03-0
Color White
Purity Grade Notes Research
Molecular Formula C10H13N5O3
Quantity 50 mg
Synonym 3'-Deoxyadenosine
Solubility Information DMSO : 50 mg/mL (199.01 mM; Need ultrasonic) ∣H2O : 2 mg/mL (7.96 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1C[C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O
Molecular Weight (g/mol) 251.24
Formula Weight 251.24
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.