missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated YES1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67196-100UL
This item is not returnable.
View return policy
Description
The YES1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YES1. This antibody recognizes human, rat antigen. The YES1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| YES1 | |
| Monoclonal | |
| 1000 μg/mL | |
| HsT441; P61-YES; Yes; c-yes | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| 62 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 1A12E7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 7525 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction