missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated TROVE2 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-67149-100UL

 View more versions of this product

Product Code. 30235474

  • ONLINE EXCLUSIVE PRICE
    £300.00
    £295.00 / 100µL
    Save £5.00
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CoraLite™ Plus 488-conjugated TROVE2 Mouse Monoclonal Antibody, Proteintechsh ows reactivity in Human, Mouse and Rat samples.
Specifications

Specifications

TROVE2
Monoclonal
1000 μg/mL
RO60; SSA2
Recombinant Protein
100 μL
Primary
Human, Mouse, Rat
60 kDa
Liquid
IgG2a
Not Available
2A4E5
CoraLite Plus 488
Mouse
Protein A purification
RUO
6738
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Conjugated Antibody
25
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.