missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated TMEM230 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | TMEM230 |
|---|---|
| Clone | 4D3A7 |
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236388
|
Proteintech
CL488-67247-100UL |
100 μL |
£309.00 £295.00 / 100µL Save £14.00 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236057
|
Proteintech
CL488-67247-20UL |
20 μL |
£124.63 £119.00 / 20µL Save £5.63 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CoraLite™ Plus 488-conjugated TMEM230 Mouse Monoclonal Antibody, Proteintechsh ows reactivity in Human, Mouse and Rat samples.Specifications
| TMEM230 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 29058 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4D3A7 | |
| Not Available | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| HSPC274; dJ1116H23.2.1 | |
| IgG2b | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title