missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated TIMELESS Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67022-20UL
This item is not returnable.
View return policy
Description
The TIMELESS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TIMELESS. This antibody recognizes human, mouse, rat antigen. The TIMELESS antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| TIMELESS | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ12640; FLJ20714; TIM; TIM1; hTIM | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 140 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 1B6G3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 8914 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu