missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Thymidylate synthase Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66725-100UL
This item is not returnable.
View return policy
Description
The Thymidylate synthase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Thymidylate synthase. This antibody recognizes human antigen. The Thymidylate synthase antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| Thymidylate synthase | |
| Monoclonal | |
| 1000 μg/mL | |
| HsT422; MGC88736; TMS; TS; TSase | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 36 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 1F5B7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7298 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering