missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated SCP3 Rabbit Polyclonal Antibody, Proteintech

Product Code. 30235555 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235555 20 μL 20µL
30236838 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235555 Supplier Proteintech Supplier No. CL4882302420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The SCP3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SCP3. This antibody recognizes human, mouse, rat antigen. The SCP3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.

Specifications

Antigen SCP3
Applications Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols COR1; MGC71888; SCP3
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50511
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 30 kDa, 33 kDa
Product Type Conjugated Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.