missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated PI3 Kinase p110 Beta Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67121-100UL
This item is not returnable.
View return policy
Description
The PI3 Kinase p110 Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Beta. This antibody recognizes human, rat antigen. The PI3 Kinase p110 Beta antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| PI3 Kinase p110 Beta | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp779K1237; MGC133043; PI3K; PI3KCB; PI3Kbeta; PIK3C1; p110-BETA | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| 120-130 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1B2A1 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5291 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction