missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated OLIG2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-13999-20UL
This item is not returnable.
View return policy
Description
The OLIG2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OLIG2. This antibody recognizes human, mouse, rat antigen. The OLIG2 antibody has been validated for the following applications: IF-P, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| OLIG2 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10215 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| BHLHB1; OLIGO2; PRKCBP2; RACK17; bHLHe19 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 32 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction