missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated OCIAD1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | OCIAD1 |
|---|---|
| Clone | 1C10C3 |
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235520
|
Proteintech
CL488-66698-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233965
|
Proteintech
CL488-66698-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The OCIAD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OCIAD1. This antibody recognizes human, mouse, rat antigen. The OCIAD1 antibody has been validated for the following applications: IF/ICC, IF-P analysis. This antibody is coralite™ plus 488.Specifications
| OCIAD1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 54940 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1C10C3 | |
| Immunocytochemistry/Immunofluorescence, Immunofluorescence | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| Asrij; FLJ20455; MGC111072; OCIA; TPA018 | |
| IgG2a | |
| Protein A purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title