missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated NEDD4L Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | NEDD4L |
|---|---|
| Clone | 2F6H6 |
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235538
|
Proteintech
CL488-67276-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235033
|
Proteintech
CL488-67276-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NEDD4L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEDD4L. This antibody recognizes human, mouse, rat antigen. The NEDD4L antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| NEDD4L | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 23327 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2F6H6 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| FLJ33870; KIAA0439; NEDD4-2; RSP5; hNedd4-2 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title