missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Myelin basic protein Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10458-20UL
This item is not returnable.
View return policy
Description
The Myelin basic protein antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Myelin basic protein. This antibody recognizes human, mouse, rat antigen. The Myelin basic protein antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| Myelin basic protein | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4155 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| MGC99675 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 14-23 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction