missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated LDHA-Specific Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-19987-20UL
This item is not returnable.
View return policy
Descripción
The LDHA-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human LDHA-Specific. This antibody recognizes human, mouse, rat antigen. The LDHA-Specific antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Especificaciones
| LDHA-Specific | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3939 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| LDH-M; LDH1; PIG19 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 32-37 kDa | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido