missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated IL-27A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66164-100UL
This item is not returnable.
View return policy
Description
The IL-27A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-27A. This antibody recognizes human antigen. The IL-27A antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| IL-27A | |
| Monoclonal | |
| 1000 μg/mL | |
| IL-27; IL-27A; IL27p28; IL30; MGC71873; p28 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 31 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 1E1H5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 246778 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction