missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated IL-22RA2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £279.00
Specifications
| Antigen | IL-22RA2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Not Available |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235947
|
Proteintech
CL488-22861-20UL |
20 μL |
£124.63 £119.00 / 20µL Save £5.63 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237706
|
Proteintech
CL488-22861-100UL |
100 μL |
£292.52 £279.00 / 100µL Save £13.52 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CoraLite™ Plus 488-conjugated IL-22RA2 Rabbit Polyclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| IL-22RA2 | |
| Not Available | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human | |
| CRF2-10; CRF2-S1; CRF2X; IL-22BP; MGC150509; MGC150510 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 116379 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title