missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated IFITM2 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-12769-20UL
This item is not returnable.
View return policy
Description
CoraLite™ Plus 488-conjugated Podoplanin Rabbit Polyclonal Antibody, Proteintech shows reactivity in Human, Mouse samples.Specifications
| IFITM2 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10581 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| 25 |
| Not Available | |
| 1000 μg/mL | |
| 1-8D | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 15 kDa | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido