missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated FSP1/S100A4 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £295.00
Specifications
| Antigen | FSP1/S100A4 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236210
|
Proteintech
CL488-16105-20UL |
20 μL |
£130.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237228
|
Proteintech
CL488-16105-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FSP1/S100A4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FSP1/S100A4. This antibody recognizes human, mouse antigen. The FSP1/S100A4 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| FSP1/S100A4 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse | |
| 18A2; 42A; CAPL; FSP1; MTS1; P9KA; PEL98 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 6275 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title