missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated F4/80 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £279.00
Specifications
| Antigen | F4/80 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235194
|
Proteintech
CL488-28463-100UL |
100 μL |
£375.00 £279.00 / 100µL Save £96.00 25% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236119
|
Proteintech
CL488-28463-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The F4/80 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse F4/80. This antibody recognizes mouse antigen. The F4/80 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| F4/80 | |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Mouse | |
| DD7A5-7; EGF-TM7; F4/80; Gpf480; Ly71; TM7LN3 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 13733 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title