missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated EIF2A/CDA02 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66482-100UL
This item is not returnable.
View return policy
Description
The EIF2A/CDA02 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF2A/CDA02. This antibody recognizes human, mouse, rat antigen. The EIF2A/CDA02 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| EIF2A/CDA02 | |
| Monoclonal | |
| 1000 μg/mL | |
| CDA02; EIF-2A; MST089; MSTP004; MSTP089 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 65 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1G7H4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 83939 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction