missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CX3CL1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60339-20UL
This item is not returnable.
View return policy
Description
CoraLite™ Plus 488-conjugated CX3CL1 Mouse Monoclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| CX3CL1 | |
| Monoclonal | |
| 1000 μg/mL | |
| ABCD-3; C3Xkine; CXC3; CXC3C; NTN; NTT; SCYD1; fractalkine; neurotactin | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 90 kDa | |
| Liquid | |
| IgG1 |
| Not Available | |
| 2F5D5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6376 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| 25 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction