missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CDC25B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | CDC25B |
|---|---|
| Clone | 1H3A4 |
| Concentration | 1000 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237505
|
Proteintech
CL488-67145-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236799
|
Proteintech
CL488-67145-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CDC25B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC25B. This antibody recognizes human, mouse, rat antigen. The CDC25B antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| CDC25B | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 994 | |
| IgG2b | |
| Protein A purification | |
| Conjugated Antibody |
| 1H3A4 | |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Mouse, Rat | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title