missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD133 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | CD133 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237065
|
Proteintech
CL488-18470-20UL |
20 μL |
£130.00 £119.00 / 20µL Save £11.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234377
|
Proteintech
CL488-18470-100UL |
100 μL |
£350.00 £295.00 / 100µL Save £55.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD133 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD133. This antibody recognizes human, mouse antigen. The CD133 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| CD133 | |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Human, Mouse | |
| AC133; CD133; CORD12; MCDR2; MSTP061; PROML1; RP41; STGD4 | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 8842 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title