missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CD107a / LAMP1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-21997-20UL
This item is not returnable.
View return policy
Description
CoraLite™ Plus 488-conjugated CD107a / LAMP1 Rabbit Polyclonal Antibody, Proteintech shows reactivity in Human samples.Specifications
| CD107a / LAMP1 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3916 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| 25 |
| Not Available | |
| 1000 μg/mL | |
| CD107a; LAMPA; LGP120 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 120 kDa | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction