missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Caspase 9/p35/p10 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66169-20UL
This item is not returnable.
View return policy
Description
The Caspase 9/p35/p10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Caspase 9/p35/p10. This antibody recognizes human, mouse antigen. The Caspase 9/p35/p10 antibody has been validated for the following applications: IF/ICC, FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| Caspase 9/p35/p10 | |
| Monoclonal | |
| 1000 μg/mL | |
| APAF-3; APAF3; CASPASE-9c; ICE-LAP6; MCH6 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 46 kDa, 35 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1B7G2 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 842 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction