missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Caspase 3/P17/P19 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-19677-100UL
This item is not returnable.
View return policy
Description
The Caspase 3/P17/P19 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Caspase 3/P17/P19. This antibody recognizes human, mouse, rat antigen. The Caspase 3/P17/P19 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| Caspase 3/P17/P19 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 836 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| 1000 μg/mL | |
| CPP32; CPP32B; SCA-1 | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 32-35 kDa, 17 kDa, 19 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction