missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Calbindin-D28k Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

£119.00 - £279.00

Specifications

Antigen Calbindin-D28k
Clone 1F8B9
Concentration 1000 μg/mL
Applications Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236126
View Documents
Proteintech
CL488-66394-20UL
20 μL
ONLINE EXCLUSIVE PRICE
£130.00
£119.00 / 20µL
Save £11.00
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30233361
View Documents
Proteintech
CL488-66394-100UL
100 μL
ONLINE EXCLUSIVE PRICE
£375.00
£279.00 / 100µL
Save £96.00
25% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Calbindin-D28k antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Calbindin-D28k. This antibody recognizes human, mouse, rat antigen. The Calbindin-D28k antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

Calbindin-D28k
1000 μg/mL
Monoclonal
Liquid
RUO
793
Recombinant Protein
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
1F8B9
Immunofluorescence
CoraLite Plus 488
Mouse
Human, Mouse, Rat
CALB
IgG1
Protein G purification
Conjugated Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.