missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated c-SRC Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60315-20UL
This item is not returnable.
View return policy
Description
The c-SRC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human c-SRC. This antibody recognizes human, hamster, mouse antigen. The c-SRC antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.Specifications
| c-SRC | |
| Monoclonal | |
| 1000 μg/mL | |
| ASV; SRC1; c-SRC; p60-Src | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Hamster, Mouse | |
| 60 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 5E10C4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6714 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction