missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated Aquaporin 4 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16473-100UL
This item is not returnable.
View return policy
Description
The Aquaporin 4 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Aquaporin 4. This antibody recognizes human, mouse, rat antigen. The Aquaporin 4 antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| Aquaporin 4 | |
| Polyclonal | |
| CoraLite Plus 488 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 361 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| HMIWC2; MGC22454; MIWC | |
| Fusion Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 35-37 kDa, 32-34 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur