missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated Annexin A2 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

£121.00 - £300.00

Specifications

Antigen Annexin A2
Clone 1C1E12
Concentration 1000 μg/mL
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233513
View Documents
Proteintech
CL488-66035-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30233732
View Documents
Proteintech
CL488-66035-100UL
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The Annexin A2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Annexin A2. This antibody recognizes human antigen. The Annexin A2 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

Annexin A2
1000 μg/mL
Monoclonal
Liquid
RUO
302
Fusion Protein
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
1C1E12
Immunocytochemistry/Immunofluorescence
CoraLite Plus 488
Mouse
Human
ANX2; ANX2L4; CAL1H; LIP2; LPC2; LPC2D; P36; PAP-IV
IgG2a
Protein A purification
Conjugated Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.