Learn More
Tocris Bioscience™ CoraFluor 1, amine reactive
Product Code. 18085122
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Product Code. Quantity unitSize
18085122 20 μg 20µg
1 options
Product Code. 18085122 Supplier Tocris Bioscience™ Supplier No. 7920/20U
Terbium cryptate FRET donor for TR-FRET assay development; amine reactive for conjugation
CoraFluor™ 1, amine reactive is a terbium-based time-resolved Förster resonance energy transfer (TR-FRET) donor for TR-FRET assay development, such as target engagement and ternary complex assays. This product is designed with a PFP ester for reactivity with primary amines, making it suitable for labeling biomolecules, including antibodies, nanobodies, proteins and peptides. It is exceptionally bright, providing unmatched assay sensitivity and stability in biological media over other commercially available TR-FRET donors. CoraFluor™ 1 amine reactive is compatible with common acceptor dyes such as FAM, TMR and Cyanine 5, SE (Cat. No. 5436), enabling seamLess assay integration in high throughput screening applications. CoraFluor™ 2, amine reactive also available. Please refer to the protocol for further information on conjugation. CoraFluor™ is a trademark of Bio-Techne Corp.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 20 μg
Target Fluorescent Probes