missing translation for 'onlineSavingsMsg'
Learn More

COR 170, Tocris Bioscience™

Product Code. 30001990
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001990

Brand: Tocris Bioscience 4756/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective CB2 receptor inverse agonist

Selective inverse agonist of CB2 receptors (Ki values are 3.8 and >10,000 nM for CB2 and CB1, respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1048039-15-1
Molecular Formula C31H36N2O2
Molecular Weight (g/mol) 468.641
InChI Key MSJISJDTJJYBFT-UHFFFAOYSA-N
Synonym n-adamant-1-yl-4-oxo-1-pentyl-6-phenyl-1,4-dihydroquinoline-3-carboxamide
PubChem CID 25033736
IUPAC Name N-(1-adamantyl)-4-oxo-1-pentyl-6-phenylquinoline-3-carboxamide
SMILES CCCCCN1C=C(C(=O)C2=C1C=CC(=C2)C3=CC=CC=C3)C(=O)NC45CC6CC(C4)CC(C6)C5

Specifications

Quantity 10 mg
Formula Weight 468.63
Percent Purity >99%
Chemical Name or Material COR 170
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.