missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COQ9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117885
This item is not returnable.
View return policy
Description
COQ9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Coenzyme Q10 (CoQ10), or ubiquinone, is a mobile lipophilic electron carrier critical for electron transfer by the mitochondrial inner membrane respiratory chain. COQ9 is 1 of several enzymes involved in biosynthesis of CoQ10 and likely functions in modification of the benzoquinone ring.
Specifications
| COQ9 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O75208 | |
| COQ9 | |
| E. coli-derived Human COQ9 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| COQ9 | |
| 2310005O14Rik; C16orf49; C78387; coenzyme Q9; coenzyme Q9 homolog; coenzyme Q9 homolog (yeast); COQ10D5; COQ9; HSPC326; PSEC0129; Ubiquinone biosynthesis protein COQ9, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57017 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction