missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COQ9 Monoclonal Antibody (K94030_7D7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555864
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: QHSETQGAEK PDPESSHSPP RYTDQGGEEE EDYESEEQLQ HRILTAALEF VPAHGWTAEA IAEGAQSLGL SSAAASMFGK DGSELILHFV TQCNTRLTRV LEEEQKLVQL GQAEKRKTDQ
Coenzyme Q10 (CoQ10), or ubiquinone, is a mobile lipophilic electron carrier critical for electron transfer by the mitochondrial inner membrane respiratory chain. COQ9 is 1 of several enzymes involved in biosynthesis of CoQ10 and likely functions in modification of the benzoquinone ring.
Specifications
| COQ9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O75208 | |
| COQ9 | |
| Recombinant human COQ9 (immunogen range is 61-180). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| K94030_7D7 | |
| Unconjugated | |
| COQ9 | |
| 2310005O14Rik; C16orf49; C78387; coenzyme Q9; coenzyme Q9 homolog; coenzyme Q9 homolog (yeast); COQ10D5; COQ9; HSPC326; PSEC0129; Ubiquinone biosynthesis protein COQ9, mitochondrial | |
| Mouse | |
| Protein A | |
| RUO | |
| 57017 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction