missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15083-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is similar to a mitochondrial di-iron containing hydroxylase in Saccharomyces cerevisiae that is involved with ubiquinone biosynthesis. Mutations in the yeast gene lead to slower development and longer life span. Alternatively spliced transcript variants have been found for this gene.Specifications
| COQ7 | |
| Polyclonal | |
| Unconjugated | |
| COQ7 | |
| CAT5, CLK 1, CLK1, COQ7, Timing protein clk 1 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10229, 12850, 25249 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97478, Q63619, Q99807 | |
| Coq7 | |
| COQ7 Fusion Protein Ag7161 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction