missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COQ5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100450
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total COQ5.
Methyltransferase required for the conversion of 2-polyprenyl-6-methoxy-1,4-benzoquinol (DDMQH2) to 2-polyprenyl-3-methyl-6-methoxy-1,4-benzoquinol (DMQH2).
Specifications
| COQ5 | |
| Polyclonal | |
| Unconjugated | |
| COQ5 | |
| 1810014G04Rik; 2-methoxy-6-polyprenyl-1,4-benzoquinol methylase, mitochondrial; coenzyme Q5 homolog, methyltransferase; coenzyme Q5 homolog, methyltransferase (S. cerevisiae); coenzyme Q5 homolog, methyltransferase (yeast); coenzyme Q5, methyltransferase; Coq5; D5Ertd33e; RGD1310857; Ubiquinone biosynthesis methyltransferase COQ5; ubiquinone biosynthesis methyltransferase COQ5, mitochondrial; Unknown (protein for MGC:142779) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 52064, 84274 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5HYK3, Q9CXI0 | |
| COQ5 | |
| A synthesized peptide derived from human COQ5(Accession Q5HYK3), corresponding to amino acid residues R123-N173. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction