missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ3 Polyclonal Antibody

COQ3 Polyclonal Antibody
Brand: Thermo Scientific PA556698
This item is not returnable.
View return policy
Description
COQ3 Polyclonal Antibody for IHC
Specifications
| COQ3 | |
| Polyclonal | |
| Unconjugated | |
| COQ3 | |
| 2-polyprenyl-6-hydroxyphenol methylase; 2-polyprenyl-6-hydroxyphenyl methylase; 3,4-dihydroxy-5-hexaprenylbenzoate methyltransferase; 3-demethylubiquinol 3-O-methyltransferase; 3-demethylubiquinone-10 3-methyltransferase; 3-demethylubiquinone-9 3-methyltransferase; 4732433J24; bA9819.1; C77934; Coenzyme Q (ubiquinone); coenzyme Q3 homolog methyltransferase; coenzyme Q3 homolog, methyltransferase; coenzyme Q3 homolog, methyltransferase (yeast); coenzyme Q3 methyltransferase; coenzyme Q3, methyltransferase; Coq3; DHHB methyltransferase; DHHBMT; DHHB-MT; DHHBMTASE; DHHB-MTase; dihydroxyhexaprenylbenzoate methyltransferase; hexaprenyldihydroxybenzoate methyltransferase, mitochondrial; methyltransferase COQ3; polyprenyldihydroxybenzoate methyltransferase; Ubiquinone biosynthesis O-methyltransferase, mitochondrial; UG0215E05 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51805 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZJ6 | |
| COQ3 | |
| Recombinant protein corresponding to Human COQ3. Recombinant protein control fragment (Product #RP-94829). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction