missing translation for 'onlineSavingsMsg'
Learn More

COQ3 Antibody (6D5), Novus Biologicals™

Product Code. 18338679 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338679 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338679 Supplier Novus Biologicals Supplier No. H00051805M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COQ3 Monoclonal antibody specifically detects COQ3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen COQ3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6D5
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_059117
Gene Alias 3-demethylubiquinone-10 3-methyltransferase, bA9819.1, coenzyme Q3 homolog, methyltransferase (S. cerevisiae), coenzyme Q3 homolog, methyltransferase (yeast), DHHB methyltransferase, DHHBMT, DHHB-MT, DHHB-MTase, DHHBMTASE, Dihydroxyhexaprenylbenzoate methyltransferase, EC 2.1.1, EC 2.1.1.114, EC 2.1.1.64, hexaprenyldihydroxybenzoate methyltransferase, mitochondrial3,4-dihydroxy-5-hexaprenylbenzoate methyltransferase, methyltransferase COQ3, UG0215E05
Host Species Mouse
Immunogen COQ3 (NP_059117, 273 a.a. ~ 369 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IVPKGTHTWEKFVSPETLESILESNGLSVQTVVGMLYNPFSGYWHWSENTSLNYAAHAVKSRVQEHPASAEFVLKGETEELQANACTNPAVHEKLKK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51805
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.