missing translation for 'onlineSavingsMsg'
Learn More

COPZ1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18360187 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360187 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360187 Supplier Novus Biologicals Supplier No. NBP310764100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COPZ1 Polyclonal specifically detects COPZ1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COPZ1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias CGI-120, coatomer protein complex, subunit zeta, coatomer protein complex, subunit zeta 1, coatomer subunit zeta-1, COPZ, Zeta-1 COP, Zeta-1-coat protein, zeta1-COP
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human COPZ1 (NP_057141). Peptide sequence DEIVDGGVILESDPQQVVHRVALRGEDVPLTEQTVSQVLQSAKEQIKWSL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22818
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.