missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101530
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total COPZ1.
This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
Specifications
| COPZ1 | |
| Polyclonal | |
| Unconjugated | |
| COPZ1 | |
| 5930435A22Rik; AA407760; CGI-120; CHUNP6876; coatomer; coatomer protein complex subunit zeta 1; coatomer protein complex, subunit zeta 1; coatomer subunit zeta-1; coatomer subunit zeta-1-like protein; COPI coat complex subunit zeta 1; COPZ; COPZ1; D4Ertd360e; HSPC181; nonclathrin coat protein zeta1-COP; Zeta-1 COP; Zeta-1-coat protein; zeta1-cop; zeta-COP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22818, 56447 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61923, P61924 | |
| COPZ1 | |
| A synthesized peptide derived from human COPZ1(Accession P61923), corresponding to amino acid residues A15-L65. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction